missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
BIOPHARMA

General-purpose swab for cleaning small and confined areas

Brand:  Berkshire™ LTO70R.20

384.02 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 12876774

  • £570.00 / Pack of 20
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polyurethane foam over rigid core
  • 100 ppi open-cell foam
  • Thermally bonded head
  • Low particles, fibers, ions and extractables, no contaminating adhesives
  • Good chemical resistance
  • Good sorbency and excellent solvent hold
  • Laundered and packaged in cleanroom

Specifications

Specifications

ISO Class 4
2.8 in.
Swab
71mm
58mm
3.8mm
12.5mm
1 in.
0.11 in.
Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials
2.8mm
2.28 in.
Polyurethane Foam
0.49 in.
3.7mm
Rigid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers