missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ Open-Top Tri-coded Tubes

Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side.

Brand:  Thermo Scientific™ 3792-WP1D-BR

782.18 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.



Additional Details : Weight : 2.25000kg

 View more versions of this product

Product Code. 17298820

  • £855.00 / Pack of 480
Estimated Shipment: 10-10-2024
to see stock.
Add to basket

Description

Description

Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side. Tubes are available in bulk packs or in barcoded latch racks. Barcoded latch racks feature a linear barcode, 2D barcode, and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific™ Matrix™ SepraSeal and DurSeal capping systems and the Thermo Scientific™ VisionMate™family of barcode readers.

Secure Tracking

  • A unique 2D barcode is laser-etched onto the base of every polypropylene open top tube to securely identify and track samples.
  • Non-proprietary 2D codes can be scanned in less than a second in VisionMate High Speed for fast decoding and sample tracking.
  • Tubes feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking

Robust Storage Rack Design

  • Matrix 2D barcoded open top storage tubes come racked in 96-format in a proprietary specially-designed, stackable, ANSI footprint.
  • Latch racks save precious space in your freezer, refrigerator, compound store or on the bench top.
  • The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop.
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to guarantee readability.
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes.
  • Every storage tube is leak tested to ensure the integrity and security of samples.

Storage Tracking Solutions for any Laboratory

  • Tubes are available with a range of sealing options including solid and pre-slit Duraseals™ and Sepraseals to meet your individual storage requirements.
  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements.
  • 2D storage tubes are compatible with Thermo Scientific freezer racks that allow optimal utilization of storage space.

Recommended for

  • Individual sample retrieval; Instant sample identification
  • Secure identification of archived samples
  • Secure shipping of samples
  • Storage of samples
  • Tracking of samples

Recycling Information

Matrix tubes, tube racks and covers are made from polypropylene copolymer and are recyclable in communities where polypropylene is recycled. Please recycle only clean, dry components that have been decontaminated of any hazardous materials or residues. The orange nylon rack latches must be removed from the tube rack base and discarded before recycling the rack base. Tip: orange latches can be removed with relative ease with the aid of pliers.

Matrix tubes are delivered in high graphic paper cartons.  Please recycle the paper cartons and product inserts.

Specifications

Specifications

Clear
Not Autoclavable
RUO
V-bottom
+25°C to -195°C
Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction
Sterile
No
Yes
2D white patch 1D linear barcode, barcoded rack
V-bottom
10 latch racks of 96 tubes/case
Yes
+77°F to -319°F
Matrix™
Yes
10 latch racks of 96 tubes/case
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers