missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
£37.31 - £325.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Concentration | 1 X |
Form | Liquid |
Product Type | Reduced-Serum Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11564506
|
Gibco™
51985026 |
500 mL |
£37.31
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11574506
|
Gibco™
51985042 |
10 x 500 mL |
£325.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology. Opti-MEM™ is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Gibco™ Opti-MEM™ modifications for a range of cell culture applications. For more information, please contact Technical Services.Gibco™ Opti-MEM™ is a unique media that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Gibco™ Opti-MEM™ with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ Opti-MEM™ is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Opti-MEM™ product made in our Grand Island facility (31985-070). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ Opti-MEM™ uses a sodium bicarbonate buffer system (2.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
1 X | |
Reduced-Serum Medium | |
Mammalian cell culture, transfection |
2 to 8°C | |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
7.0 to 7.4 |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
For In Vitro Diagnostic Use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title