Learn More
Gibco™ Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement

Description
Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.
This Opti-MEM™ I is modified as follows:.
With: GlutaMAX™, Phenol Red.
The complete formulation is confidential. For more information, please contact Technical Services.
Using Opti-MEM™ I Medium.
Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Line | Sp2, AE-1, CHO, BHK-21, and HEK |
| Cell Type | Primary Fibroblasts |
| Classification | Animal Origin |
| Concentration | 1X |
| Form | Liquid |
| Product Type | Opti-MEM I |
| Sterility | Sterile-filtered |
| With Additives | GlutaMAX, Phenol Red |
| Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
| Product Line | Gibco, GlutaMAX, Opti-MEM |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.