Learn More
Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles
Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles.
£178.00 - £4070.00
Specifications
Certifications/Compliance | Includes Certificate of Analysis |
---|---|
Content And Storage | 2°C to 8°C |
Description | Opti-Bind Polystyrene Sulfate Particles |
For Use With (Application) | Diagnostic Assays |
Material | Polystyrene Sulfate |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
11930085
|
Thermo Scientific™
71002697100250 |
1.25 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11940085
|
Thermo Scientific™
71002697100350 |
1.25 μm | 100 mL |
£585.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11899912
|
Thermo Scientific™
71003497100250 |
2.5 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11809922
|
Thermo Scientific™
71003497100350 |
2.5 μm | 100 mL |
£585.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11804153
|
Thermo Scientific™
81000397100290 |
0.1 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11814153
|
Thermo Scientific™
81000397100390 |
0.1 μm | 100 mL |
£585.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11824153
|
Thermo Scientific™
81000597100290 |
0.2 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11834153
|
Thermo Scientific™
81000597100390 |
0.2 μm | 100 mL |
£585.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11844153
|
Thermo Scientific™
81000797100290 |
0.3 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11854153
|
Thermo Scientific™
81000797100390 |
0.3 μm | 100 mL |
£585.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11864153
|
Thermo Scientific™
81000997100290 |
0.4 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11874153
|
Thermo Scientific™
81000997100390 |
0.4 μm | 100 mL |
£675.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11874183
|
Thermo Scientific™
91001397100290 |
0.6 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11884183
|
Thermo Scientific™
91001397100390 |
0.6 μm | 100 mL |
£585.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11894183
|
Thermo Scientific™
91001897100290 |
0.85 μm | 15 mL |
£178.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11804193
|
Thermo Scientific™
91001897100390 |
0.85 μm | 100 mL |
£740.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11814193
|
Thermo Scientific™
91001897100490 |
0.85 μm | 1000 mL |
£4070.00
1000mL |
Estimated Shipment: 06-12-2024
Log in to see stock. |
||||
Description
Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles for use in turbidimetric/nephelometric assays, slide agglutination assays, chemiluminescent assays and in clinical diagnostics and molecular biology applications. Particles can be used direct from the bottle, without any pre-cleaning.
Our production does not utilize common surfactants (like SDS, Tween™ 20, Triton™ X-100) that can interfere with protein binding to particle surfaces. Polystyrene surfaces are very hydrophobic and adsorb proteins almost instantaneously. Our proprietary anionic surfactant does not interfere with the binding of proteins or cause proteins to desorb from microparticle surfaces.- Available in a wide range of diameters (from 0.1 μM to 2.5 μM)
- No pre-washing
- Tight size distribution—CV<2%
- Optimized colloidal stability
- Shelf-life of 10 years
Recommended for:
Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Bind™ particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.
Specifications
Includes Certificate of Analysis | |
Opti-Bind Polystyrene Sulfate Particles | |
Polystyrene Sulfate | |
Package Insert and Certificate of Analysis |
2°C to 8°C | |
Diagnostic Assays | |
Specialty Particles | |
Opti-Bind Polystyrene particles are available in different concentrations and package sizes upon request. |
Safety and Handling
missing translation for 'shelfLife' : 10 Years