missing translation for 'onlineSavingsMsg'
Learn More

Sherwood Scientific™ Optical Gelatin Filter

Brand:  Sherwood Scientific™ 25221001

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12722685

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

± 20 nm
Optical Gelatin Filter
1 Ea.
600 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers