missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sherwood Scientific™ Optical Gelatin Filter
Brand: Sherwood Scientific™ 25216001
Code : 03
Additional Details : Weight : 1.00000kg
Specifications
±20 nm | |
Optical Gelatin Filter | |
1 Ea. | |
470 nm |