Learn More
BRAND™ Optical Glass Cavity Slides
With rectangular ground edges.
Brand: BRAND™ 475535
Code : A0
Additional Details : Weight : 0.80000kg
Description
- White (clear) soda-lime glass, hydrolytic class 3
- Single concavity
- Provided in packs of 50
Specifications
Ground | |
Soda Lime Glass | |
1.2 to 1.5 mm | |
Clear White |
Optical Glass Slide | |
2 | |
50 |