missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Optifit Twin™ Full-Face Air Purifying Respirator (APR)

The Optifit Twin™ Full-Face APR provides protection versus particulates and/or toxic gases. It features a full lens for wide field of vision, a silicone body for optimum fit, and a click-fit cartridge system for flexible protection. A built-in diaphragm allows for better speech transmission.

£215.00 - £278.00

Specifications

Cartridge/Filter Bayonet N-Series filter (EN 141 or EN 143 Class 1 or 2 cert Honeywell filters only)
Certifications/Compliance EN 136:1998, EC Category PPE: 3, QA: ISO 9001/2000, EC Certificate: 0070-236-079-06-06-0023, EC Attestation: 0070-236-079-06-06-0023, Edeclaration of conformity INRS - 1715231
Color Black
Product Type Full-Face Air Purifying Respirator (APR)
With Exhalation Valve Yes
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11856930
View Documents
Honeywell™
1715241
Medium
£278.00
Each
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
 
11867000
View Documents
Honeywell™
1715251
Large
£215.00
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
 
12535924
View Documents
Honeywell™
1715231
Small
£235.00
Each
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
 
Description

Description

  • Product Type: Full-Face Air Purifying Respirator (APR)
  • Description: Polycarbonate Visor with Silicone Skirt, Face Blank, and Exhalation Valve. Integrated Plastic Diaphragm
  • Cartridge/Filter: Bayonet N-Series Filter (EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters Only)
  • Lens Type: Full Lens
  • Material (Lens): Polycarbonate
  • Harness Style: Five-Strap Head Harness, with Quick Release Adjusting Buckles
  • Certifications/Compliance: EN136:1998, CE PPE Cat:3, QA: ISO 9001/2000
  • For Use With (Application): Laboratories, Pharmaceutical Industry, Oil and Gas Industry, Mechanical Industry, Automotive Industry, Chemical Industry
Specifications

Specifications

Bayonet N-Series filter (EN 141 or EN 143 Class 1 or 2 cert Honeywell filters only)
Black
Yes
Laboratories, Pharmaceutical Industry, Oil and Gas Industry, Mechanical Industry, Automotive Industry, Chemical Industry
Polycarbonate
EN 136:1998, EC Category PPE: 3, QA: ISO 9001/2000, EC Certificate: 0070-236-079-06-06-0023, EC Attestation: 0070-236-079-06-06-0023, Edeclaration of conformity INRS - 1715231
Full-Face Air Purifying Respirator (APR)
Silicone
Five-strap head harness, with quick release adjusting buckles
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell™ Optifit Twin™ Full-Face Air Purifying Respirator (APR)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.