Learn More
Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.
£191.00 - £5220.00
Specifications
Additive Proprietary anionic surfactant
Certifications/Compliance Includes Certificate of Analysis
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
For Use With (Application) Covalent Coupling
View More Specs
Products 30
Product Code Brand Quantity Diameter (Metric) Surface Acid Loading Concentration Price Quantity & Availability  
Product Code Brand Quantity Diameter (Metric) Surface Acid Loading Concentration Price Quantity & Availability  
11819922
View Documents
Thermo Scientific™
73003305100250
15 mL 2 μm High Acid 10%
£201.00
15mL
11829922
View Documents
Thermo Scientific™
73003305100350
100 mL 2 μm High Acid 10%
£757.00
100mL
11950085
View Documents
Thermo Scientific™
73003420100250
15 mL 3 μm High Acid 10%
£218.00
15mL
11960085
View Documents
Thermo Scientific™
73003420100350
100 mL 3 μm High Acid 10%
£757.00
100mL
12406462
View Documents
Thermo Scientific™
83000520100290
15 mL 0.2 μm High Acid 10%
£201.00
15mL
12416462
View Documents
Thermo Scientific™
83000520100390
100 mL 0.2 μm High Acid 10%
£757.00
100mL
12612550
View Documents
Thermo Scientific™
83000550100290
15 mL 0.2 μm Medium Acid 10%
£201.00
15mL
12446462
View Documents
Thermo Scientific™
83000550100390
100 mL 0.2 μm Medium Acid 10%
£757.00
100mL
11976558
View Documents
Thermo Scientific™
83000550100390
100 mL 0.2 μm Medium Acid 10% This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11894153
View Documents
Thermo Scientific™
83000720100290
15 mL 0.3 μm High Acid 10%
£201.00
15mL
11804163
View Documents
Thermo Scientific™
83000720100390
100 mL 0.3 μm High Acid 10%
£757.00
100mL
11814163
View Documents
Thermo Scientific™
83000750100290
15 mL 0.3 μm Medium Acid 10%
£201.00
15mL
11824163
View Documents
Thermo Scientific™
83000750100390
100 mL 0.3 μm Medium Acid 10%
£757.00
100mL
11834163
View Documents
Thermo Scientific™
83000920100290
15 mL 0.4 μm High Acid 10%
£201.00
15mL
11844163
View Documents
Thermo Scientific™
83000920100390
100 mL 0.4 μm High Acid 10%
£757.00
100mL
11894163
View Documents
Thermo Scientific™
83000970100290
15 mL 0.4 μm Low Acid 10%
£201.00
15mL
11804173
View Documents
Thermo Scientific™
83000970100390
100 mL 0.4 μm Low Acid 10%
£757.00
100mL
11814173
View Documents
Thermo Scientific™
83000970100490
1000 mL 0.4 μm Low Acid 10%
£5220.00
1000mL
11844203
View Documents
Thermo Scientific™
93000570100290
15 mL 0.2 μm Low Acid 10%
£201.00
15mL
11854203
View Documents
Thermo Scientific™
93000570100390
100 mL 0.2 μm Low Acid 10%
£803.00
100mL
11990075
View Documents
Thermo Scientific™
W050CA
15 mL 0.5 μm Medium Acid 4%
£213.00
15mL
11900085
View Documents
Thermo Scientific™
W050CB
100 mL 0.5 μm Medium Acid 4%
£722.00
100mL
11910085
View Documents
Thermo Scientific™
W080CA
15 mL 0.85 μm Medium Acid 4%
£191.00
15mL
11920085
View Documents
Thermo Scientific™
W080CB
100 mL 0.85 μm Medium Acid 4%
£722.00
100mL
11804962
View Documents
Thermo Scientific™
W090CA
15 mL 0.9 μm High Acid 4%
£191.00
15mL
12493072
View Documents
Thermo Scientific™
W090CB
100 mL 0.9 μm High Acid 4%
£722.00
100mL
11814962
View Documents
Thermo Scientific™
W400CA
15 mL 4.0 μm High Acid 4%
£201.00
15mL
12433082
View Documents
Thermo Scientific™
W400CB
100 mL 4.0 μm High Acid 4%
£757.00
100mL
11824962
View Documents
Thermo Scientific™
W500CA
15 mL 5.0 μm High Acid 4%
£201.00
15mL
12453082
View Documents
Thermo Scientific™
W500CB
100 mL 5.0 μm High Acid 4%
£1420.00
100mL
Description
Description
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.
  • Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization
  • Surface area per gram of a particle varies inversely with the particle diameter
  • Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based
  • For most applications, particles can be used directly from the bottle without any pre-cleaning
  • Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces or cause proteins to desorb from particle surfaces
  • Various surface acid concentrations available to help optimize reagent development efforts
  • Particles have been used in and are equally successful in molecular biology and research applications
Parking Area Properties
  • Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
  • Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
  • High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling
Recommended for:
  • Clinical diagnostics
  • Molecular biology
  • Nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.
Specifications
Specifications
Proprietary anionic surfactant
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Covalent Coupling
Specialty Particles
1.05g/cm³
Includes Certificate of Analysis
Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
Polystyrene Sulfate
Package Insert Sheet containing Certicate of Analysis
Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request.