Learn More
Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column
POA - £1075.00
Specifications
Stationary Phase | Optima™ 5 |
---|---|
USP Type | G27 |
Type | High Performance GC Capillary Column |
For Use With (Application) | Gas Chromatography (GC) |
Temperature Range | 340°C |
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Film Thickness | Price | Quantity & Availability | |||
10621302
|
Macherey-Nagel™
726056.10 |
0.25 mm | 10 m | 0.25 μm |
£235.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10236561
|
Macherey-Nagel™
726523.10 |
0.53 mm | 10 m | 0.5 μm |
£292.00
Pack of 10 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10746702
|
Macherey-Nagel™
726541.10 |
0.53 mm | 10 m | 1 μm |
£292.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10337971
|
Macherey-Nagel™
726525.10 |
0.53 mm | 10 m | 2 μm |
£292.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10206231
|
Macherey-Nagel™
726541.15 |
0.53 mm | 15 m | 1 μm |
£352.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10775932
|
Macherey-Nagel™
726056.15 |
0.25 mm | 15 m | 0.25 μm |
£315.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10630732
|
Macherey-Nagel™
726916.10 |
0.53 mm | 10 m | 5 μm |
£292.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10266371
|
Macherey-Nagel™
726911.25 |
0.25 mm | 25 m | 0.1 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10002861
|
Macherey-Nagel™
726056.25 |
0.25 mm | 25 m | 0.25 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10650342
|
Macherey-Nagel™
726623.25 |
0.25 mm | 25 m | 0.35 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10092901
|
Macherey-Nagel™
726099.25 |
0.25 mm | 25 m | 0.5 μm |
£524.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10144991
|
Macherey-Nagel™
726807.25 |
0.25 mm | 25 m | 1 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10620352
|
Macherey-Nagel™
726854.25 |
0.2 mm | 25 m | 0.1 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10286421
|
Macherey-Nagel™
726857.25 |
0.2 mm | 25 m | 0.2 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10155091
|
Macherey-Nagel™
726860.25 |
0.2 mm | 25 m | 0.35 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10786142
|
Macherey-Nagel™
726863.25 |
0.2 mm | 25 m | 0.5 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10690912
|
Macherey-Nagel™
726523.25 |
0.53 mm | 25 m | 0.5 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10194791
|
Macherey-Nagel™
726541.25 |
0.53 mm | 25 m | 1 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10631302
|
Macherey-Nagel™
726525.25 |
0.53 mm | 25 m | 2 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10105091
|
Macherey-Nagel™
726916.25 |
0.53 mm | 25 m | 5 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10649961
|
Macherey-Nagel™
726911.30 |
0.25 mm | 30 m | 0.1 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10609001
|
Macherey-Nagel™
726056.30 |
0.25 mm | 30 m | 0.25 μm |
£510.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10378591
|
Macherey-Nagel™
726623.30 |
0.25 mm | 30 m | 0.35 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10777082
|
Macherey-Nagel™
726099.30 |
0.25 mm | 30 m | 0.5 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10699571
|
Macherey-Nagel™
726807.30 |
0.25 mm | 30 m | 1 μm |
£510.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10022871
|
Macherey-Nagel™
726523.30 |
0.53 mm | 30 m | 0.5 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10378301
|
Macherey-Nagel™
726541.30 |
0.53 mm | 30 m | 1 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10546681
|
Macherey-Nagel™
726525.30 |
0.53 mm | 30 m | 2 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10461472
|
Macherey-Nagel™
726916.30 |
0.53 mm | 30 m | 5 μm |
£630.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10062991
|
Macherey-Nagel™
726911.50 |
0.25 mm | 50 m | 0.1 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10739752
|
Macherey-Nagel™
726056.50 |
0.25 mm | 50 m | 0.25 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10165131
|
Macherey-Nagel™
726623.50 |
0.25 mm | 50 m | 0.35 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10536511
|
Macherey-Nagel™
726099.50 |
0.25 mm | 50 m | 0.5 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10715962
|
Macherey-Nagel™
726860.50 |
0.2 mm | 50 m | 0.35 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10194941
|
Macherey-Nagel™
726857.50 |
0.2 mm | 50 m | 0.2 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10144801
|
Macherey-Nagel™
726807.50 |
0.25 mm | 50 m | 1 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10063051
|
Macherey-Nagel™
726863.50 |
0.2 mm | 50 m | 0.5 μm |
£845.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10206851
|
Macherey-Nagel™
726525.50 |
0.53 mm | 50 m | 2 μm |
£975.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10266611
|
Macherey-Nagel™
726916.50 |
0.53 mm | 50 m | 5 μm |
POA
Each |
N/A | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
10690722
|
Macherey-Nagel™
726623.60 |
0.25 mm | 60 m | 0.35 μm |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10460892
|
Macherey-Nagel™
726056.60 |
0.25 mm | 60 m | 0.25 μm |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10556681
|
Macherey-Nagel™
726099.60 |
0.25 mm | 60 m | 0.5 μm |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10216851
|
Macherey-Nagel™
726525.60 |
0.53 mm | 60 m | 2 μm |
£1075.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10104851
|
Macherey-Nagel™
726807.60 |
0.25 mm | 60 m | 1 μm |
£880.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
Specifications
Optima™ 5 | |
High Performance GC Capillary Column | |
340°C |
G27 | |
Gas Chromatography (GC) | |
1 Each |