Learn More
Macherey-Nagel™ Optima™ 5 MS Accent GC Capillary Column
£370.00 - £1300.00
Specifications
Stationary Phase | Optima™ 5 MS Accent |
---|---|
USP Type | G1, G2, G38 |
For Use With (Equipment) | Ideal for Ion Trap and Qadrupole MS Detectors |
Type | GC Capillary Column |
For Use With (Application) | Gas Chromatography (GC) |
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Film Thickness | Price | Quantity & Availability | |||
12422830
|
Macherey-Nagel™
725815.12 |
0.2 mm | 12 m | 0.35 μm |
£370.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12442830
|
Macherey-Nagel™
725820.15 |
0.25 mm | 15 m | 0.25 μm |
£444.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12452820
|
Macherey-Nagel™
725810.25 |
0.2 mm | 25 m | 0.2 μm |
£655.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12492820
|
Macherey-Nagel™
725812.25 |
0.32 mm | 25 m | 1 μm |
£705.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12452830
|
Macherey-Nagel™
725820.30 |
0.25 mm | 30 m | 0.25 μm |
£655.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12472830
|
Macherey-Nagel™
725825.30 |
0.25 mm | 30 m | 0.5 μm |
£655.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12472820
|
Macherey-Nagel™
725811.30 |
0.32 mm | 30 m | 0.25 μm |
£705.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12492830
|
Macherey-Nagel™
725826.30 |
0.25 mm | 30 m | 1 μm |
£655.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12412830
|
Macherey-Nagel™
725813.30 |
0.32 mm | 30 m | 0.5 μm |
£705.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12462820
|
Macherey-Nagel™
725810.50 |
0.2 mm | 50 m | 0.2 μm |
£1110.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12432830
|
Macherey-Nagel™
725815.50 |
0.2 mm | 50 m | 0.35 μm |
£1110.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12462830
|
Macherey-Nagel™
725820.60 |
0.25 mm | 60 m | 0.25 μm |
£1175.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12482830
|
Macherey-Nagel™
725825.60 |
0.25 mm | 60 m | 0.5 μm |
£1175.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12402840
|
Macherey-Nagel™
725826.60 |
0.25 mm | 60 m | 1 μm |
£1175.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12482820
|
Macherey-Nagel™
725811.60 |
0.32 mm | 60 m | 0.25 μm |
£1300.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
12402830
|
Macherey-Nagel™
725812.60 |
0.32 mm | 60 m | 1 μm |
£1300.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
Especificaciones
Optima™ 5 MS Accent | |
Ideal for Ion Trap and Qadrupole MS Detectors | |
Gas Chromatography (GC) | |
1 Each |
G1, G2, G38 | |
GC Capillary Column | |
320°C to 340°C |