Learn More
Macherey-Nagel™ Optima™ 5 MS GC Capillary Column
Brand:  Macherey-Nagel™ 726225.60
 View more versions of this product
Product Code. 10536331
Specifications
0.4 mm
0.25 mm
G27, G36
320°C to 340°C
60 m
GC Capillary Column
Optima™ 5 MS
GC/MS, ECD
0.5 μm
1 Each