Learn More
Macherey-Nagel™ Optima™ 5 Performance GC Capillary Column
£249.00 - £970.00
Specifications
Diameter (Metric) | 0.5 mm |
---|---|
Diameter (Metric) Inner | 0.32 mm |
Stationary Phase | Optima™ 5 |
USP Type | G27 |
Type | High Performance GC Capillary Column |
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Film Thickness | Price | Quantity & Availability | |||
10421472
|
Macherey-Nagel™
726313.10 |
0.32 mm | 10 m | 0.1 μm |
£249.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10368351
|
Macherey-Nagel™
726313.15 |
0.32 mm | 15 m | 0.1 μm |
£322.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10629961
|
Macherey-Nagel™
726314.15 |
0.32 mm | 15 m | 0.25 μm |
£322.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10516161
|
Macherey-Nagel™
726325.15 |
0.32 mm | 15 m | 1 μm |
£322.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10194701
|
Macherey-Nagel™
726934.15 |
0.32 mm | 15 m | 5 μm |
£322.00
Pack of 15 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
12897361
|
Macherey-Nagel™
726934.15 |
0.32 mm | 15 m | 5 μm |
£322.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10266321
|
Macherey-Nagel™
726313.25 |
0.32 mm | 25 m | 0.1 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10556861
|
Macherey-Nagel™
726628.25 |
0.32 mm | 25 m | 0.35 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10236421
|
Macherey-Nagel™
726314.25 |
0.32 mm | 25 m | 0.25 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10459551
|
Macherey-Nagel™
726316.25 |
0.32 mm | 25 m | 0.5 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10490312
|
Macherey-Nagel™
726325.25 |
0.32 mm | 25 m | 1 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10577041
|
Macherey-Nagel™
726809.25 |
0.32 mm | 25 m | 3 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10104711
|
Macherey-Nagel™
726934.25 |
0.32 mm | 25 m | 5 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10378351
|
Macherey-Nagel™
726313.30 |
0.32 mm | 30 m | 0.1 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10195031
|
Macherey-Nagel™
726314.30 |
0.32 mm | 30 m | 0.25 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10728432
|
Macherey-Nagel™
726628.30 |
0.32 mm | 30 m | 0.35 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10053001
|
Macherey-Nagel™
726316.30 |
0.32 mm | 30 m | 0.5 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10680912
|
Macherey-Nagel™
726325.30 |
0.32 mm | 30 m | 1 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10776132
|
Macherey-Nagel™
726809.30 |
0.32 mm | 30 m | 3 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10134941
|
Macherey-Nagel™
726313.50 |
0.32 mm | 50 m | 0.1 μm |
£915.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10308171
|
Macherey-Nagel™
726934.30 |
0.32 mm | 30 m | 5 μm |
£545.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10378541
|
Macherey-Nagel™
726314.50 |
0.32 mm | 50 m | 0.25 μm |
£915.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10489741
|
Macherey-Nagel™
726628.50 |
0.32 mm | 50 m | 0.35 μm |
£915.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10012871
|
Macherey-Nagel™
726325.50 |
0.32 mm | 50 m | 1 μm |
£915.00
Pack of 50 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10195081
|
Macherey-Nagel™
726316.50 |
0.32 mm | 50 m | 0.5 μm |
£915.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10042991
|
Macherey-Nagel™
726809.50 |
0.32 mm | 50 m | 3 μm |
£915.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10092961
|
Macherey-Nagel™
726313.60 |
0.32 mm | 60 m | 0.1 μm |
£970.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10640152
|
Macherey-Nagel™
726314.60 |
0.32 mm | 60 m | 0.25 μm |
£970.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10641112
|
Macherey-Nagel™
726628.60 |
0.32 mm | 60 m | 0.35 μm |
£970.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10216661
|
Macherey-Nagel™
726316.60 |
0.32 mm | 60 m | 0.5 μm |
£970.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10194551
|
Macherey-Nagel™
726325.60 |
0.32 mm | 60 m | 1 μm |
£970.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
10052991
|
Macherey-Nagel™
726809.60 |
0.32 mm | 60 m | 3 μm |
£970.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
Specifications
0.5 mm | |
Optima™ 5 | |
High Performance GC Capillary Column | |
340°C |
0.32 mm | |
G27 | |
Gas Chromatography (GC) | |
1 Each |