missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ OptiPRO™ SFM
GREENER_CHOICE

Product Code. 11530426
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11530426

Brand: Gibco™ 12309019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production.

OptiPRO™ SFM is a serum-free, animal origin-free culture medium designed for growth of several kidney-derived cell lines including MDBK, MDCK, PK-15 and VERO for virus or recombinant protein production. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7. Gibco™ OptiPRO™ SFM features:

  • Serum-free, animal origin-free, low protein formulation
  • Direct adaptation for many popular cell lines

Serum-free, animal origin-free, low protein formulation

Gibco™ OptiPRO™ SFM is formulated with a very low protein concentration (< 10 μg/mL), allowing for easier purification of your recombinant protein.

Direct adaptation for many popular cell lines

Gibco™ OptiPRO™ SFM has been designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO and PK-15. In addition, OptiPRO™ SFM has successfully grown other cell lines, such as Hela, BHK-21 and COS-7, allowing you to choose a single medium for multiple cell lines.

cGMP Manufacturing and Quality System

Gibco™ OptiPRO™ SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, MDCK, BHK-21, PK-15, COS-7, MDBK, VERO
Classification Animal Origin-free, Low Protein, Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium
Serum Level Serum-free
With Additives Phenol Red
Without Additives No Glutamine
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle(s)
Included Antibiotics No antibiotics
Content And Storage Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.