Learn More
OPTIZYME™ 10X Buffer 4, Fisher BioReagents™
Brand: Fisher Bioreagents BP8087-1
Code : 32
Additional Details : Weight : 0.01700kg
Description
1X Buffer Composition:
33mM Tris-acetate (pH 7.9 at 37°C), 10mM magnesium acetate, 66mM potassium acetate, 0.1mg/mL BSA
Specifications
33mM Tris Acetate (pH 7.9 at 37°C), 10mM Magnesium Acetate, 66mM Potassium Acetate, 0.1mg/mL BSA | |
5 x 1 mL | |
Liquid |
-20°C | |
Buffer 4 |