Learn More
OPTIZYME™ 10X Buffer BamHI, Fisher BioReagents™
Brand:  Fisher BioReagents BP8089-1
Product Code. 11944862
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 8.0 at 37°C), 5mM MgCl2, 100mM KCl , 0.02 % Triton X-100, 0.1mg/mL BSA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10mM Tris HCl (pH 8 at 37°C), 5mM MgCl2, 100mM KCl, 0.02 % Triton™ X-100, 0.1mg/mL BSA
5 x 1 mL
Liquid
-20°C
Buffer BamHI
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • OPTIZYME
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OPTIZYME™ 10X Buffer BamHI, Fisher BioReagents™ > 5 x 1mL