Learn More
OPTIZYME™ 10X Buffer KpnI, Fisher BioReagents™
Brand:  Fisher BioReagents BP8094-1
Product Code. 11994862
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.02% Triton X-100 and 0.1mg/mL BSA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10mM Tris HCl (pH 7.5 at 37°C), 10mM MgCl2, Triton™ X 100 0.02%, 0.1mg/mL BSA
1 mL
Liquid
-20°C
10X Buffer KpnI
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • OPTIZYME
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OPTIZYME™ 10X Buffer KpnI, Fisher BioReagents™ > 1mL