Learn More
OPTIZYME™ 10X Buffer Sau3AI, Fisher BioReagents™
Brand:  Fisher BioReagents BP8091-1
Product Code. 11954862
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
1X Buffer Composition:
33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate, 0.02% Triton X-100 and 0.1mg/mL BSA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
33mM Tris Acetate (pH 7.9 at 37°C), 10mM Magnesium Acetate, 66mM Potassium Acetate, 0.1mg/mL BSA, 0.02% Triton™ X-100
1 mL
Liquid
-20°C
Sau3AI
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • OPTIZYME
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OPTIZYME™ 10X Buffer Sau3AI, Fisher BioReagents™ > Qty.: 1mL