missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ M-MLV Reverse Transcriptase, Fisher BioReagents™

£57.32 - £129.33

Specifications

Concentration 200 U/μL
Color Colorless
pH 8.3
For Use With (Application) Generation of labeled cDNA probes for microarrays, synthesis of cDNA for cloning and expression, analysis of RNA by primer extension, DNA labeling, optimum activity at 42°C, incorporates modified Nucleotides
Storage Buffer OPTIZYME™ M-MLV RT Buffer 5X : Tris HCl 250mM (8.3 pH at 25°C), KCl 250mM, MgCl2 20mM, DTT 50mM
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12827162
View Documents Promotion Details
Fisher BioReagents
BP8104-1
5000 U This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12837162
View Documents Promotion Details
Fisher BioReagents
BP8104-5
25,000 U This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

OPTIZYME™ M-MuLV RT is a genetically modified MLV RT that possesses RNA- and DNA-dependent polymerase activity.

Applications and Features:

  • OPTIZYME M-MLV RT is used for efficient first strand cDNA synthesis for RT-PCR and real-time RT-PCR (up to 13kb)
  • Generation of labeled cDNA probes for microarrays
  • Synthesis of cDNA for cloning and expression
  • Analysis of RNA by primer extension
  • DNA labeling
  • Optimum activity at 42°C (active up to 50°C)
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labeled nucleotides)

Supplied with:
5X OPTIZYME M-MLV RT Buffer: 250mM Tris-HCl (pH 8.3 at 25°C), 250mM KCl, 20mM MgCl2, 50mM DTT
Source: E. Coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus reverse transcriptase
Quality Control:

  • Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free
  • Functionally tested in first strand cDNA and RT-PCR

Specifications

Specifications

200 U/μL
8.3
OPTIZYME™ M-MLV RT Buffer 5X : Tris HCl 250mM (8.3 pH at 25°C), KCl 250mM, MgCl2 20mM, DTT 50mM
E. coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus Reverse Transcriptase
Liquid
Colorless
Generation of labeled cDNA probes for microarrays, synthesis of cDNA for cloning and expression, analysis of RNA by primer extension, DNA labeling, optimum activity at 42°C, incorporates modified Nucleotides
-20°C
OPTIZYME™ M-MLV Reverse Transcriptase
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • OPTIZYME

SDS
Documents

Documents

Product Certifications