missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ opus™ Dispenser
Guarantees excellent accuracy during serial dispensing. Hirschmann Instruments™ opus™ Dispenser is for motor-driven dispensing directly from the reagent bottle.
Brand: Hirschmann™ 9581050
This item is not returnable.
View return policy
Description
- Available in three different volumes
- Dispenser touch screen module incl. brown UV protection
- Complete electronic control
- With reagent recirculation system
- Serial and USB interface
Specifications
0.6% | |
50 mL | |
230 V |
Opus Dispenser | |
For motor-powered dispensing directly from the reagent bottle | |
50 Hz |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hirschmann™ opus™ Dispenser > Capacity: 50mL
Spot an opportunity for improvement?Share a Content Correction