Learn More
Thermo Scientific™ Oral-Eze™ Collection Devices
Take the guesswork out of collecting oral fluid samples with innovative indicator window technology.
£1775.00
Specifications
Detectable Analytes | None |
---|---|
Sample Type | Oral Fluid |
Type | Collection Device |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
17825878
|
Thermo Scientific™
96100-500 |
Oral-Eze Collection Device | 500 |
£1775.00
Pack of 500 |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Simplify the collection of oral samples for routine drug testing with our innovative oral fluid collection system. The Oral-Eze™ Collection Device available from Thermo Scientific™ provides all the advantages of previous collection systems with the added benefit of our indicator window technology - taking the guesswork out of sample collection.
Using the Oral-Eze Oral Fluid Collection System simplifies the collection of oral samples for routine drug testing.- Collection system and drug tests used with it are FDA-cleared
- Unique indicator window turns blue when complete, taking the guesswork out of the process and reducing repeat collections
- Fast collection time is typically 3 to 5 minutes
- Specimen and any drugs in the specimen are stable for 21 days after collection
- Non-negative specimens tested by the laboratory are stable for at least one year when stored frozen
Specifications
None | |
Oral Fluid | |
Collection Device |