missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Organochlorine Pesticides QC Sample, Wastewater Standard, SPEX CertiPrep™
Shop alle SPEX Certiprep produkter
Klik for at se tilgængelige muligheder
Quantity:
2 mL
Pakningsstørrelse:
2mL
Beskrivelse
Indeholder
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Organochlorine Pesticide Standards
- Wastewater analysis
- 2 mL volume
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Tekniske data
| Chemical Name or Material | Organochlorine Pesticides QC Sample |
| Name Note | Yields up to 2L after dilution |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Industry Type | Wastewater Testing |
| Quantity | 2 mL |
Product intended for laboratory use only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion