missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organometallic Oil Standard, 12-Element, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£163.00 - £253.00
Specifications
Chemical Name or Material | 12 Element Oil Standard |
---|---|
Packaging | Triple-leached LDPE bottle |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11470339
|
SPEX Certiprep
S12-100Z |
50 g | Triple-leached LDPE bottle |
£163.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11460339
|
SPEX Certiprep
S12-100Y |
100 g | Triple-leached LDPE bottle |
£253.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 100μg/g |
Al | 100μg/g |
Cr | 100μg/g |
Cu | 100μg/g |
Fe | 100μg/g |
Mg | 100μg/g |
Na | 100μg/g |
Ni | 100μg/g |
Pb | 100μg/g |
Si | 100μg/g |
Sn | 100μg/g |
Ti | 100μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
12 Element Oil Standard | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Each component at 100μg/g (S12-100), 300μg/g (S12-300), 500μg/g (S12-500), or 900μg/g (S12-900) in Base Oil 75: Aluminum Chromium Copper Iron Lead Magnesium Nickel Silicon Silver Sodium Tin Titanium |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Oil | |
Base Oil 75 |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only