missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organometallic Oil Standard, 21-Element, 300μg/g, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£148.00 - £256.00
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11440349
|
SPEX Certiprep
S21-300Z |
50 g | Triple-leached LDPE bottle |
£148.00
50g |
Estimated Shipment: 29-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11430349
|
SPEX Certiprep
S21-300Y |
100 g | Triple-leached LDPE bottle |
£256.00
100g |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 300μg/g |
Al | 300μg/g |
B | 300μg/g |
Ba | 300μg/g |
Ca | 300μg/g |
Cd | 300μg/g |
Cr | 300μg/g |
Cu | 300μg/g |
Fe | 300μg/g |
Mg | 300μg/g |
Mn | 300μg/g |
Mo | 300μg/g |
Na | 300μg/g |
Ni | 300μg/g |
P | 300μg/g |
Pb | 300μg/g |
Si | 300μg/g |
Sn | 300μg/g |
Ti | 300μg/g |
V | 300μg/g |
Zn | 300μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Product intended for laboratory use only