Learn More
Organometallic Oil Standard, 21-Element For Wear Metal Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep S21-900Z
Code : 04
Additional Details : Weight : 0.09000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 900μg/g |
Al | 900μg/g |
B | 900μg/g |
Ba | 900μg/g |
Ca | 900μg/g |
Cd | 900μg/g |
Cr | 900μg/g |
Cu | 900μg/g |
Fe | 900μg/g |
Mg | 900μg/g |
Mn | 900μg/g |
Mo | 900μg/g |
Na | 900μg/g |
Ni | 900μg/g |
P | 900μg/g |
Pb | 900μg/g |
Si | 900μg/g |
Sn | 900μg/g |
Ti | 900μg/g |
V | 900μg/g |
Zn | 900μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
21 Element Oil Standard | |
50 g | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Oil |
Triple-leached LDPE bottle | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Base Oil 75 |
Product intended for laboratory use only