Learn More
Thermo Scientific™ Orion™ 8102BNUWP ROSS Ultra™ pH Electrode
Conduct precise pH determinations with this general purpose, top performance glass body combination electrode with waterproof BNC connector.
Brand:  Thermo Scientific™ 8102BNUWP
Product Code. 10544973
  • £309.37 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Conduct precise pH determinations with the Thermo Scientific™ Orion™ 8102BNUWP ROSS Ultra™ pH Electrode, a general purpose, top performance glass body combination electrode with waterproof BNC connector. The pH response is independent of temperature changes and can be used on any pH meter with a BNC connection.
Includes:
  • 60ml bottle of Orion ROSS electrode fill solution, 810007
Requires:
  • pH buffers for calibration
  • Beakers, plastic or glass
  • Magnetic stirrer or Orion stirrer probe
  • Distilled or deionized water
Compatible with:
Waterproof BNC connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.
Recommended for:
  • Q.C. and research applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass body combination electrode
Ceramic
0 to 14
pH Electrode
3M KCl
ROSS
32°F to 212°F
1 m
WPBNC
Glass Body
0°C to 100°C
<0.002pH/day
60ml bottle of Orion ROSS electrode fill solution
0.01
39.4 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products