Learn More
Thermo Scientific™ Orion™ 900100 Sure-Flow™ Reference Half-Cell Electrode
Precisely measure with ion-selective electrodes using this sleeve-type Ag/AgCl reference electrode.
Brand:  Thermo Scientific™ 900100
Product Code. 10101021
  • £205.12 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Precisely measure with ion-selective electrodes using the Thermo Scientific™ Orion™ 900100 Sure-Flow™ Reference Half-Cell Electrode, a sleeve-type Ag/AgCl reference electrode.
Ordering Information:
Reference electrode Orion 900100 is recommended for all pH and ion-selective measurements except for those:
  • In dilute solutions where the ion to be measured is contained in the filling solution of the Orion 900100
  • In solutions where an ion in the filling solution would introduce an electrode interference
  • In solutions where a species is present which forms a precipitate with an ion contained in the filling solution of the Orion 900100
For these samples, the double junction reference electrode, Orion 900200, with an appropriate intermediate filling solution, is recommended.
When filled with Orion 900001 Filling Solution, the Orion 900100 matches the potential characteristics of a conventional KCl saturated calomel electrode.
Recommended for:
  • Many ISE determinations
  • Routine pH measurements with Orion 910100 Electrodes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
900100 Sure-Flow Reference Half Cell Electrode
0 to 14
Reference Half-Cell Electrode
Orion 900001 Filling Solution
Ag/AgCl
110 mm
Sure-Flow Sleeve Junction
0°C to 100°C
Single Junction Ref Electrode
ISE determinations
Equitransferent solution with Ag/AgCI
12 mm