missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Double Junction Semi-Micro Stem Glass Bodied Combination pH Electrode
Click to view available options
Connector Types:
BNC (Waterproof)
Unit Size:
Each
Description
Thermo Scientific™ Orion™ Double Junction Semi-Micro Electrodes provide reliable and accurate measurements of routine samples. They are available in chemically resistant glass bodies and best suited for measuring general samples as small as 0.2mL, in test tubes and small containers.
Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.
Solutions required
Electrode fill solution
910008 3M KCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
910008 3M KCl filling solution, 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Specifications
| Description | Double Junction pH Electrode |
| Connector Types | BNC (Waterproof) |
| Junction | Ceramic |
| Material | Glass Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 60°C |
| Type | Double Junction pH Electrode |
| Filling Solution Type | 3M KCl |
| Internal Reference | Ag/AgCl Double Junction |
| Precision | 0.02 |
| Show More |
Documents
Scientific Resources
Application Notes
Titratable Acidity of Red Wine by Manual Titration (Potentiometric)
File size: 127.4 kB
Measuring pH in Wine and Juice
File size: 155.3 kB
Measuring pH of Concentrated Samples
File size: 416.4 kB
Measuring pH in Low Ionic Strength Solutions
File size: 177.7 kB
Product Information
Manuals
AquaPro and Double Junction pH Electrodes
Version: FEB.2016
File size: 158.7 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Orion™ Double Junction Semi-Micro Stem Glass Bodied Combination pH Electrode >
Spot an opportunity for improvement?Share a Content Correction