missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Glass Bodied Temperature Probes

Product Code. p-4529705
Click to view available options
Connector Types:
3.5 mm Phono Jack
8-pin DIN
Mini-DIN
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10685091

Brand: Thermo Scientific™ 917006

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Automatically correct pH measurements for variation in electrode slope due to temperature change with this glass ATC probe.

Automatically correct pH measurements for variation in electrode slope due to temperature change with the Thermo Scientific™ Orion™ 927006MD Glass ATC Probe. Available with three different connector options.

Note: Three different models are available to accommodate different Orion meters. MiniDIN version is compatible with older meters with use of an adapter cable.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±1°C
For Use With (Equipment) Orion Star A Meters
Material Glass
Temperature (Metric) Operating 0°C to 100°C
Type ATC Probe
pH Range 0 to 14
Diameter (English) Body 0.3 in.
Diameter (Metric) Body 8 mm
Length (English) 4.7 in.
Length (Metric) 120 mm
Temperature (English) Operating 32°F to 212°F
Connector Types 8-pin DIN
Storage Requirements Store in air
Show More Show Less
compliance-icons
Product Identifier
  • QConnect Yellow
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-3(2H)-isothiazolone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.