Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ3170 Colorimeter
Gather accurate chlorine measurements with a colorimeter designed for both the laboratory and the field in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
Brand:  Thermo Scientific™ CLSK200
144.69 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.02600kg
 View more versions of this product
Product Code. 15994002
  • £152.31 / Each
Estimated Shipment: 01-11-2024
to see stock.
Description
Description
The Thermo Scientific™ Orion™ AQUAfast™ AQ3170 colorimeter has a rugged and waterproof design to provide accurate chlorine measurements whether testing in the laboratory or the field. This colorimeter is designed to improve the user experience by including a large, backlit display, a real time clock and automatic data logging. The AQUAfast AQ3170 colorimeter is used in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
  • Large digital backlit display
  • Automatic switch-off
  • User friendly keypad
  • Real time clock and date
  • Calibration mode
  • Storage function
  • Waterproof sample chamber
  • Angled, ergonomic design for easy bench top or hand-held use
  • Suppression of chloride interference
  • Ability to export data to a PC
  • ST-COD tubes ready for use
  • 2-year warranty
Specifications
Specifications
AQUAfast chlorine secondary standards (0.0, 0.20 and 1.0 mg/L chlorine) for AQ3170 colorimeter
AQ3170 Chlorine Colorimeter
Sealed vials with 0.0, 0.2 and 1.0 mg/L secondary chlorine standards and case