missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ AQ4000 Colorimeter and Accessories

Measure common water and wastewater parameters using this colorimeter with 65 pre-programmed methods and auto-test ampoule compatibility.

Brand:  Thermo Scientific™ AQ4000

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11956069

  • £1187.47 / Each
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

Measure common colorimetric water and wastewater parameters using the Thermo Scientific™ Orion™ AQUAfast AQ4000 Colorimeter. This instrument has 65 pre-programmed methods, automatic selection of four wavelengths and timer. Use auto-test ampoules with Auto-ID barcodes to simplify sample preparation and tests. Ampoules contain pre-measured liquid reagent and fill with the exact volume of sample. The colorimeter reads the barcode and automatically identifies the parameter and initiates the test. This IP67 waterproof meter records 100 data points and offers an average battery life of 2,500 hours intermittent use.

Features:

  • Identifies the species to be measured
  • Ensures error-free readings at a very affordable price
  • Selects the program and initiates the measurement without the user ever having pushed a button
  • Guides user through the setup and measurement with simple operation
  • Waterproof to IP67 standards

Benefits:

  • User can customize calibration and create completely custom method
  • Up to 12 data points can be used to define custom method
  • User can store maximum of 10 methods in the colorimeter's memory
  • User can datalog 100 points in the field with a time and date tag

Ordering Information:

For use with Orion™ AQUAfast™ AQ4000, Orion™ AQUAfast™ AQ3070, Orion™ AQUAfast™ AQ3700 and discontinued Orion AQ2 series meters.

Note: Uses Thermo Scientific™ Auto-ID™ ampoules, liquid, powder and tablet reagent including many methods accepted for EPA, ISO and other regulatory reporting.

Specifications

Specifications

0 to 2 A
13, 16, 24 mm
AA
IP67
420, 520, 580, 610 nm
90 at 50°C max. %
Keypad 12 dual function keys with tactile feedback
±2 nm
Up to 190 and 10 user defined programmed methods
2500 hr (Alkaline); 10000 hr (Lithium)
Colorimeter for use with 189 pre-programmed method
Custom LCD
LED
Automatic
AQ4000 Colorimeter, 13 and 16 mm vial adapter, tamping rod, 16 mm vial, zero kit (AQ4ZER), 4 AA Batteries and carrying case
100
0°C to 45°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.