Learn More
Thermo Scientific™ Orion™ AquaMate™ Power Cords
Product Code. p-4569694
Click to view available options
Voltage:
220 V
Plug Type:
EU Plug
Unit Size:
Each
Product Code. 15189182
Brand: Thermo Scientific™ AQ220V
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Thermo Scientific™ Orion™ AquaMate Power Cords with AquaMate 8000 UV-Vis and AquaMate 7000 Vis spectrophotometers
Use Thermo Scientific™ Orion™ AquaMate Power Cords with AquaMate 8000 UV-Vis and AquaMate 7000 Vis spectrophotometers. Select power cords with a US / Japan plug, Euro plug, UK / Singapore plug or China / Australia / New Zealand plug.
AquaMate Power Cords include the AquaMate 110V US, Japan Plug Power Cord (Cat. No. AQ110V) and AquaMate 220V Euro Plug Power Cord (Cat. No. AQ220V). The AquaMate 240V UK, Singapore Plug Power Cord (Cat. No. AQ240V) and AquaMate China, Australia, New Zealand Plug Power Cord (Cat. No. AQCHNV). Purchase separately.
TRUSTED_SUSTAINABILITY
Specifications
Product Line AquaMate
Type Power Cord
Voltage 220 V
Plug Type EU Plug
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.