Learn More
Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results
Brand: Thermo Scientific™ AQ8100
6531.53 GBP valid until 2025-12-31
Use promo code "24356" to get your promotional price.
Includes: Test Tube Holder for vials and test tube from 12 to 25 mm OD, 10 mm square single-cell holder, AquaMate Water Analysis Methods Software, Snap-on Printer, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362)
Description
Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.
Pre-programmed Methods:
- More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
- Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
- Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries
Versatility:
- Glove friendly 7-inch touch screen controller
- Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD
Verification:
- Performance verification tests ensure wavelength accuracy and instrument functionality
- Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
- Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
- Built-in test methods for stray light, noise and resolution are available to validate instrument performance further
Compliance
CE / TUV
Services
1 Year

Specifications
±0.002 A | |
Accessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down | |
35.5 cm | |
Dual Silicon Photodiodes | |
35.5 x 38.5 x 19.5 cm | |
UV-Vis Spectrophotometer | |
<0.0005 A/hr. (At 500 nm after 1 hour warmup) | |
Water and Waste Water Analysis | |
Test Tube Holder for vials and test tube from 12 to 25 mm OD, 10 mm square single-cell holder, AquaMate Water Analysis Methods Software, Snap-on Printer, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362). | |
Glove- friendly touchscreen | |
35.5 cm | |
Double Beam | |
-3A to +5A | |
±0.001A at 1A measured at 1.0A at 546 nm | |
Slow, medium and fast - up to 1600 nm/min. | |
1 Year | |
0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm | |
7.5 kg |
CE / TUV | |
Single USB-A Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available Print via USB, Ethernet or Wi-Fi USB adaptor available |
|
AquaMate 8100 UV-Vis Spectrophotometer with North America, European and UK Power cord | |
7 in. High Definition Color Touchscreen, Fixed, 800 x 1280 pixels | |
2 nm | |
190 to 1100 nm | |
External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz | |
19.5 cm | |
Glove friendly touch-screen | |
>5 years typical | |
(RMS at 500 nm 60 consecutive measurements) ≤0.00020A at 0A at 260 and 500 nm ≤0.00030A at 1A at 260 and 500 nm ≤0.00040A at 2A at 260 and 500 nm | |
±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A | |
-2A to +3.5A | |
Snap-on printer included | |
< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2) | |
±0.5 nm | |
< ±0.2 nm | |
38.5 cm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.