missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes

Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions by using these pH combination electrodes.

Brand:  Thermo Scientific™ 9107APMD

 View more versions of this product

Product Code. 10809610

  • £241.80 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions with Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes. These electrodes, which have a clog-resistant and open double junction reference system, are ideal for measurements in samples containing proteins or TRIS buffers and that do not contain mercury. The electrodes require minimal maintenance because there is no fill solution; instead they have a polymer gel reference.

Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.
  • Measure 0 to 14pH
  • Temperature range: 0° to 60°C (0° to 140°F)
  • Choice of connectors with 1m (3ft.) cable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epoxy Body pH/ATC
Open
0 to 14
Combination Ag/AgCl double juction pH Electrode, Mini-DIN Connector
Orion pH Meter
±0.02
32°F to 140°F
39 in.
1 m
12 mm
BNC Waterproof, 8-pin MiniDIN
Epoxy Body
0°C to 60°C
General Purpose
Ag/AgCl Double Junction
Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
4.72 in.
120 mm
0.47 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers