missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes
Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions by using these pH combination electrodes.
Brand: Thermo Scientific™ 9115APWP
This item is not returnable.
View return policy
Description
Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions with Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes. These electrodes, which have a clog-resistant and open double junction reference system, are ideal for measurements in samples containing proteins or TRIS buffers and that do not contain mercury. The electrodes require minimal maintenance because there is no fill solution; instead they have a polymer gel reference.
Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.- Measure 0 to 14pH
- Temperature range: 0° to 60°C (0° to 140°F)
- Choice of connectors with 1m (3ft.) cable
Specifications
Epoxy Body | |
Open | |
0 to 14 | |
Combination pH Electrode | |
Ag/AgCl Double Junction | |
Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with. | |
6.5 in. | |
165 mm | |
0.32 in. |
WPBNC | |
Epoxy Body | |
0°C to 60°C | |
Small sample sizes, test tubes, aqueous solutions only | |
±0.02 | |
32°F to 140°F | |
39 in. | |
1 m | |
8 mm |