Learn More
Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler Accessories
Maintain automated sample analysis with accessories for our legacy autosampler.
Brand: Thermo Scientific™ ASB050
Description
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler. With a choice of 48- and 28-position racks, this products offers flexibility with interchangeable racks to accommodate sample size and volume.
- Interchangeable racks to accommodate sample size and volume
- Choice of 48- and 28-position racks
- Beaker sizes of 45-, 50- and 150-mL
- Replacement electrode holders for each model
Specifications
50 mL beakers | |
Titrator Accessories | |
AT5050 AutoTration-500 Autosampler with 48-position rack |
Autosampler 50 mL plastic beakers, 50 pack | |
50 beakers (50 mL) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.