Learn More
Thermo Scientific™ Orion™ Conductivity Standards and Solutions
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Brand:  Thermo Scientific™ 01100710
 View more versions of this product
Product Code. 11707817
  • £42.85 / Pack of 10
Explore more special offers
Description
Description
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conductivity Standard
Pouch
Standard Solution
Yes
Orion pH Meter
100°C
Soluble
692 ppm as NaCl
10 x 15 mL
Clear
Conductivity Standard
4.75 to 7.75
1413 μS/cm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Hazard Statement
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.