Learn More
Thermo Scientific™ Orion™ Conductivity Standards and Solutions
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Brand: Thermo Scientific™ 01100910
40.18 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature

Specifications
Conductivity Standard | |
Pouch | |
Standard Solution | |
Yes | |
Orion pH Meter | |
100°C | |
Soluble |
— | |
10 x 15 mL | |
Clear | |
Conductivity Standard | |
4.75 to 7.75 | |
147 μS/cm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.