missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Conductivity Standards and Solutions

Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.

Brand:  Thermo Scientific™ 990106

78.08 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11747717

  • £83.54 / 475mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.

Conductivity Standards

  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Conductivity Standard
475 mL
12.9 mS
Clear
100°C
Soluble
Conductivity Standard
Bottle
Yes
0.1M KCl
4.75 to 7.75
Orion pH Meter
Standard Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCR1 Certificate TypeCertificate of Analysis Date30/12/2024 Catalog Number990106
Lot NumberCX1 Certificate TypeCertificate of Analysis Date07/03/2024 Catalog Number990106
Lot NumberBQ1 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number990106
Lot NumberBW1 Certificate TypeCertificate of Analysis Date26/05/2023 Catalog Number990106
Lot NumberAV1 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number990106
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    27

    Hours

    12

    Minutes

    38

    Seconds

    40
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    58

    Hours

    12

    Minutes

    38

    Seconds

    40
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ Conductivity Standards and Solutions > 0.1M KCl Conductivity Standard, 475mL Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.