Learn More
Thermo Scientific™ Orion™ Cupric Electrodes
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically.
Brand: Thermo Scientific™ 9429SC
678.02 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Measure cupric ions in aqueous solutions quickly, simply, accurately and economically with the Thermo Scientific™ Orion™ Cupric Electrodes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.- Calibration Standards Catalog number: 0.1 M Cu(NO3)2/942906
- Required ISA Catalog number: 940011
Specifications
Screw Cap | |
Free-flowing liquid junction assures stable, drift-free potentials | |
Combination Ion Selective Electrode |
With screw cap connector | |
ISE Meters |