Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Brand: Thermo Scientific™ 090035
This item is not returnable.
View return policy
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
- For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
- Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Especificaciones
BNC | |
Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors | |
LEMO Miniature (Mettler) |
LEMO Miniature (Mettler); BNC; BNC to LEMO Miniature Adapter | |
Electrode Adapter Cable |