missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Electrode Adapter Cables

Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.

£79.33 - £197.49


Products 2
Product Code Brand Connector Types Meter Input Price Quantity & Availability  
Product Code Brand Connector Types Meter Input Price Quantity & Availability  
10274442
View Documents Promotion Details
Thermo Scientific™
91CBNC
Screw Cap BNC
£79.33
Each
In Stock
to see stock.
Add to basket
 
11722867
View Documents
Thermo Scientific™
91USRF
Screw Cap 2 mm Pin Tip
£197.49
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
 
Description

Description

Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.

  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Warranty: Out of Box
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ Electrode Adapter Cables

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.