Learn More
Thermo Scientific™ Orion™ Filling Solutions for Process Analyzer Reference Electrodes
Enhance the performance and extend the life of ion selective electrodes used in our process analyzers with this filling solution.
Brand:  Thermo Scientific™ 181073
Code : NEW
Additional Details : CAS Number : 9002-93-1 Weight : 0.50000kg
 View more versions of this product
Product Code. 15827134
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Enhance the performance and extend the life of ion selective electrodes used in Orion™ process analyzers with Thermo Scientific™ Orion™ Filling Solutions for Process Analyzer Reference Electrodes.
Filling solutions for Orion process analyzer reference electrodes
Specifications
Specifications
Filling Solution for Orion Process Analyzer 100056/210056 Reference Electrodes, 5 x 60mL
Filling Solution
Orion™ 100056/210056 Reference Electrodes
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Reference Electrode Internal Filling Solution
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cesium chloride, triton x-100
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.