Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
£71.88 - £214.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10408532
|
Thermo Scientific™
950204 |
Carbon Dioxide Electrode 9502BNMP |
£183.93
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
10292441
|
Thermo Scientific™
951204 |
Standard Ammonia Electrode 9512BNWP, 9512 |
£124.60
Pack of 20 |
In Stock
Log in to see stock. |
|||||
10758971
|
Thermo Scientific™
951205 |
Standard Ammonia Electrode 9512BNWP, 9512 |
£164.43
Pack of 3 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
10554454
|
Thermo Scientific™
951214 |
High Performance Ammonia Electrode 9512HPBNWP, 9512HP |
£113.58
Pack of 20 |
In Stock
Log in to see stock. |
|||||
10165673
|
Thermo Scientific™
951215 |
High Performance Ammonia Electrode 9512HPBNWP |
£71.88
Pack of 3 |
In Stock
Log in to see stock. |
|||||
11762178
|
Thermo Scientific™
954604 |
Nitrogen Oxide Electrode | N/A | ||||||
Description
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
Specifications
Gas-Sensing ISE Membrane |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.