Learn More
Thermo Scientific™ Orion™ Industrial Ion Selective Electrodes
Ensure accurate results for your most important measurements with Thermo Scientific™ Orion™ Industrial Ion Selective Electrodes.
Brand:  Thermo Scientific™ LTR100049
 View more versions of this product
Product Code. 10125502
  • £537.68 / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure accurate results for your most important measurements with Thermo Scientific™ Orion™ Industrial Ion Selective Electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calcium Sensing Half-Cell Electrode
Ion Selective Electrode
Certificate of Analysis (COA), Certificate of Conformance (COC)
2120XP, 1820AO Analyzers