Learn More
Thermo Scientific™ Orion™ Iodide Electrodes
Get fast and stable readings using these iodide electrodes.
Brand:  Thermo Scientific™ 9453BN
Code : 59
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10169860
  • £644.51 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
Get fast and stable readings using Thermo Scientific™ Orion™ Iodide Electrodes with built-in Thermo Scientific™ Sure-Flow™ reference. Combination design decreases the amount of required sample and reduces waste.
  • Calibration standards catalog number: 945306 - 0.1M NaI Required ISA catalog number: 940011
Specifications
Specifications
BNC
With BNC connector
ISE Meters
Solid state half-cell