Learn More
Thermo Scientific™ Orion™ Ion Specific Electrode Kits
Ensure correct measurement by using high quality electrodes and reagents for ion specific measurement. Ordering is easy! All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis
£713.34 - £985.00
Specifications
Connector Types | BNC, waterproof |
---|---|
Length (English) | 4.75 in. |
Length (English) Cable | 39.40 in. |
Length (Metric) Cable | 1m |
Material | Epoxy |
Product Code | Brand | Description | Applications | Electrode | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Applications | Electrode | Price | Quantity & Availability | |||
15839656
|
Thermo Scientific™
9609BNLSLN |
Low Level Fluoride Ion Selective Electrode and Reagent Kit | Fluoride Analysis | Fluoride ISE |
£713.34
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
17742719
|
Thermo Scientific™
9512BNSLN |
STD Ammonia Ion Selective Electrode and Reagent Kit | Ammonia Analysis | Ammonia ISE |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16661624
|
Thermo Scientific™
9512HPBNSLN |
HP Ammonia Ion Selective Electrode and Reagent Kit | Ammonia Analysis | HP Ammonia ISE |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Perform quick and accurate measurements by using high quality Thermo Scientific™ Orion™ Ion Specific Electrodes and reagents for ion specific measurement. Ordering is easy! We offer a comprehensive portfolio of ion selective electrode calibration standards, ionic strength adjusters, reagents, and fill solutions for your ISE measurement needs. All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis.
- Orion Combination Fluoride ISE features the Thermo™ Scientific™ Orion™ Sure-Flow™ junction which contributes to highly stable and precise fluoride measurements while simplifying electrode maintenance.
- 0.5, 1.0, 2.0, 5.0 and 10.0 ppm Fluoride Standards are pre-mixed with total ionic strength adjustor for your convenience. No dilution and no addition of ionic strength adjustor needed.
- Total Ionic Strength Adjustor (TISAB II) included for addition to samples
- This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
- The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
- This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
- The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
Specifications
BNC, waterproof | |
39.40 in. | |
Epoxy | |
ISE Meter |
4.75 in. | |
1m | |
Kit | |
25°C |