missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents

Product Code. 11767687
Click to view available options
Type:
Acid Buffer
Acid Reagent
Alkaline Reagent
CISA
ISA Solution
Iodide Reagent
Ionic Strength Adjustor Buffer
Nitrate Interference Suppressor
Reconditioning Solution
Storage Solution
Sulfide Anti-Oxidant Buffer
TISAB II
TISAB III
pH Adjusting ISA
pH Adjusting Low Level ISA
For Use With (Equipment):
Ammonia Electrode
Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode
Calcium Electrode
Carbon Dioxide Electrode
Chloride Electrode
Chlorine Electrode
Cyanide Electrode
Fluoride Electrode
Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode
Nitrate Electrode
Nitrogen Oxide Electrode
Potassium Electrode
Sodium Electrode
Sodium ISE
Sulfide Electrode
Quantity:
475 mL
5 x 50 mL
1 Gal.
4 x 475 mL
2 x 1 L
4 x 1 Gal.
Unit Size:
3.8 L
475mL
Each
Pack of 2
Pack of 4
Pack of 5
This item is not returnable. View return policy

Product Code. 11767687

Brand: Thermo Scientific™ 956410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

Warranty: Out of box

Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.

All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:

  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
TRUSTED_SUSTAINABILITY

Specifications

Description Acid Buffer for Nitrogen Oxide Electrode
For Use With (Equipment) Nitrogen Oxide Electrode
Type Acid Buffer
For Use With (Application) Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Packaging Bottle
Quantity 475 mL
compliance-icons
Product Identifier
  • Acid Buffer
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
Supplemental information
  • MIXTURE LIST-Contains : potassium nitrate, sulphuric acid

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.