Learn More
Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Description
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
Warranty: Out of box
Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.
All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:
- Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
- Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
- Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
- Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
Specifications
Specifications
| Description | pH Adjusting ISA for Ammonia Electrode |
| For Use With (Equipment) | Ammonia Electrode |
| Type | pH Adjusting ISA |
| For Use With (Application) | Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes |
| Packaging | Bottle |
| Quantity | 475 mL |
Safety and Handling
- Ammonia pH - Adjusting ISA
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H314-Causes severe skin burns and eye damage.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- H318-Causes serious eye damage.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P321-Specific treatment (see on this label).
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P314-Get medical advice/attention if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sodium Hydroxide, Disodium EDTA, Methanol
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.